Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013055.1 | NP_001013073.1 | 3 - 4 | 402 | 125 | |
XM_006253790.1 | XP_006253852.1 | 3 - 4 | 364 | 125 | ||
GenBank mRNA | BC091191.1 | - | 3 - 4 | 402 | 7368273 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |