Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014008.1 | NP_001014030.1 | 1003 | 122 | ||
XM_006253695.2 | XP_006253757.1 | 2873 | 122 | |||
GenBank mRNA | BC083711.1 | - | 1003 | 7190620 | 122 |
Gene Symbol: NEWGENE_1549776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253707.2 | XP_006253769.1 | 7 - 8 | 2873 | 122 | |
XM_008771557.1 | XP_008769779.1 | 6 - 7 | 2949 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |