Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082409.1 | NP_001075878.1 | 6 - 7 | 795 | 83 | |
XM_008771539.1 | XP_008769761.1 | 6 - 7 | 840 | 83 | ||
XM_008771540.1 | XP_008769762.1 | 6 - 7 | 840 | 83 | ||
XM_008771541.1 | XP_008769763.1 | 6 - 7 | 840 | 83 | ||
XM_008771542.1 | XP_008769764.1 | 6 - 7 | 1802 | 83 | ||
GenBank mRNA | BC167744.1 | - | 6 - 7 | 783 | 7099140 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |