Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013157.1 | NP_001013175.1 | 5 - 6 | 828 | 61 | |
XM_006231968.2 | XP_006232030.1 | 5 - 6 | 950 | 61 | ||
XM_006231969.2 | XP_006232031.1 | 5 - 6 | 955 | 61 | ||
XM_006231970.2 | XP_006232032.1 | 5 - 6 | 802 | 61 | ||
XM_006231971.2 | XP_006232033.1 | 5 - 6 | 827 | 61 | ||
XM_006231972.2 | XP_006232034.1 | 5 - 6 | 890 | 61 | ||
XM_008760774.1 | XP_008758996.1 | 5 - 6 | 871 | 61 | ||
GenBank mRNA | BC091271.1 | - | 5 - 6 | 828 | 7381610 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |