Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106874.1 | NP_001100344.1 | 6 - 7 | 816 | 74 | |
XM_006244305.1 | XP_006244367.1 | 6 - 7 | 865 | 74 | ||
XM_008766754.1 | XP_008764976.1 | 6 - 7 | 926 | 74 | ||
GenBank mRNA | FM165287.1 | - | 6 - 7 | 753 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |