Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fibcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107829.1 | NP_001101299.1 | 3 - 4 | 892 | 87 |
Gene Symbol: NEWGENE_1309097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761689.1 | XP_008759911.1 | 3 - 4 | 868 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |