Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031662.1 | NP_001026832.1 | 3 - 4 | 303 | 75 | |
GenBank mRNA | BC099123.1 | - | 3 - 4 | 303 | 7462864 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |