Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234024.2 | XP_006234086.1 | 3 - 4 | 625 | 94 | |
GenBank mRNA | AY169317.1 | - | 1 - 2 | 121 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Inventoried Probe spans exons |