Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033868.1 | NP_001029040.1 | 3 - 4 | 434 | 66 | |
GenBank mRNA | CK358924.1 | - | 3 - 4 | 368 | 66 |
Gene Symbol: LOC102553670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233722.2 | XP_006233784.1 | 2 - 3 | 2286 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |