Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107176.1 | NP_001100646.1 | 2 - 3 | 533 | 102 | |
XM_006249871.2 | XP_006249933.1 | 3 - 4 | 1380 | 102 | ||
XM_006249872.2 | XP_006249934.1 | 3 - 4 | 1939 | 102 | ||
XM_008769554.1 | XP_008767776.1 | 2 - 3 | 1766 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |