Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017494.1 | NP_001017494.1 | 2 - 3 | 186 | 117 | |
GenBank mRNA | BC079114.1 | - | 2 - 3 | 186 | 7126053 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |