Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250013.2 | XP_006250075.1 | 2 - 3 | 469 | 68 | |
XM_008769664.1 | XP_008767886.1 | 2 - 3 | 466 | 68 | ||
GenBank mRNA | JN864041.1 | - | 2 - 3 | 256 | 68 | |
JN864042.1 | - | 2 - 3 | 256 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |