Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048042.1 | NP_001041507.1 | 2 - 3 | 305 | 105 | |
XM_008769614.1 | XP_008767836.1 | 2 - 3 | 163 | 105 | ||
GenBank mRNA | DQ022370.1 | - | 2 - 3 | 305 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |