Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100974.1 | NP_001094444.1 | 9 - 10 | 1465 | 106 | |
XM_006254807.2 | XP_006254869.1 | 9 - 10 | 1524 | 106 | ||
XM_006254808.2 | XP_006254870.1 | 9 - 10 | 3464 | 106 | ||
XM_006254809.2 | XP_006254871.1 | 9 - 10 | 1431 | 106 | ||
XM_006254811.2 | XP_006254873.1 | 9 - 10 | 1560 | 106 | ||
GenBank mRNA | AB199794.1 | - | 9 - 10 | 1465 | 106 | |
BC168236.1 | - | 1507 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |