Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fech | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108434.1 | NP_001101904.1 | 4 - 5 | 557 | 77 | |
XM_006254850.2 | XP_006254912.1 | 4 - 5 | 2811 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |