Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107386.1 | NP_001100856.1 | 1 - 2 | 584 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |