Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107386.1 | NP_001100856.1 | 3 - 4 | 824 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |