Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035002.1 | NP_001030174.1 | 8 - 9 | 873 | 97 | |
XM_008760330.1 | XP_008758552.1 | 8 - 9 | 958 | 97 | ||
GenBank mRNA | BC094514.1 | - | 8 - 9 | 873 | 7307334 | 97 |
EF688601.1 | - | 7 - 8 | 688 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |