Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231111.2 | XP_006231173.1 | 4 - 5 | 527 | 58 | |
XM_008760278.1 | XP_008758500.1 | 2 - 3 | 233 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |