Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006973.1 | NP_001006974.1 | 4 - 5 | 513 | 60 | |
XM_006230739.2 | XP_006230801.1 | 4 - 5 | 557 | 60 | ||
GenBank mRNA | BC083740.1 | - | 4 - 5 | 513 | 7191782 | 60 |
FQ225172.1 | - | 4 - 5 | 538 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons |