Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108097.3 | NP_001101567.2 | 1 - 2 | 155 | 106 | |
XM_006241381.2 | XP_006241443.1 | 1 - 2 | 172 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |