Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271073.1 | NP_001258002.1 | 1 - 2 | 60 | 72 | |
NM_001271074.1 | NP_001258003.1 | 1 - 2 | 60 | 72 | ||
NM_001271075.1 | NP_001258004.1 | 1 - 2 | 60 | 72 | ||
NM_001271076.1 | NP_001258005.1 | 1 - 2 | 60 | 72 | ||
NM_001271077.1 | NP_001258006.1 | 1 - 2 | 60 | 72 | ||
NM_001271078.1 | NP_001258007.1 | 1 - 2 | 60 | 72 | ||
GenBank mRNA | U51195.1 | - | 1 - 2 | 61 | 72 | |
U60562.1 | - | 1 - 2 | 61 | 72 | ||
U60564.1 | - | 1 - 2 | 56 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |