Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271073.1 | NP_001258002.1 | 3 - 4 | 169 | 72 | |
NM_001271074.1 | NP_001258003.1 | 3 - 4 | 169 | 72 | ||
NM_001271075.1 | NP_001258004.1 | 3 - 4 | 169 | 72 | ||
NM_001271076.1 | NP_001258005.1 | 3 - 4 | 169 | 72 | ||
NM_001271077.1 | NP_001258006.1 | 3 - 4 | 169 | 72 | ||
NM_001271078.1 | NP_001258007.1 | 3 - 4 | 169 | 72 | ||
GenBank mRNA | U01245.2 | - | 2 - 3 | 128 | 72 | |
U07054.1 | - | 2 - 3 | 110 | 72 | ||
U51195.1 | - | 3 - 4 | 170 | 72 | ||
U60562.1 | - | 3 - 4 | 170 | 72 | ||
U60563.1 | - | 1 - 2 | 50 | 72 | ||
U60564.1 | - | 3 - 4 | 165 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |