Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014111.1 | NP_001014133.1 | 10 - 11 | 1317 | 147 | |
XM_006256835.2 | XP_006256897.2 | 11 - 12 | 1718 | 147 | ||
XM_006256836.2 | XP_006256898.2 | 11 - 12 | 1706 | 147 | ||
XM_006256839.2 | XP_006256901.2 | 11 - 12 | 1703 | 147 | ||
XM_008773169.1 | XP_008771391.1 | 11 - 12 | 1721 | 147 | ||
GenBank mRNA | BC086548.1 | - | 10 - 11 | 1317 | 7301795 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |