Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037187.1 | NP_001032264.1 | 1 - 2 | 113 | 131 | |
XM_006256890.2 | XP_006256952.1 | 1 - 2 | 173 | 131 | ||
XM_008773168.1 | XP_008771390.1 | 1 - 2 | 173 | 131 | ||
GenBank mRNA | BC099110.1 | - | 1 - 2 | 113 | 7461546 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |