Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109503.1 | NP_001102973.1 | 2 - 3 | 208 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |