Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebna1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008721.1 | NP_001008721.1 | 786 | 76 | ||
GenBank mRNA | BC087738.1 | - | 786 | 7310934 | 76 | |
FQ213511.1 | - | 809 | 76 | |||
FQ219880.1 | - | 731 | 76 | |||
FQ231568.1 | - | 808 | 76 |
Gene Symbol: LOC100912375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750007.3 | XP_003750055.1 | 7 - 8 | 806 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |