Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108112.1 | NP_001101582.1 | 12 - 13 | 1511 | 73 | |
XM_006257400.2 | XP_006257462.1 | 12 - 13 | 1458 | 73 | ||
GenBank mRNA | BC166473.1 | - | 12 - 13 | 1411 | 9090730 | 73 |
FQ231766.1 | - | 4 - 5 | 548 | 73 |
Gene Symbol: LOC103690014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765882.1 | XP_008764104.1 | 10 - 11 | 1087 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |