Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013222.1 | NP_001013240.1 | 4 - 5 | 551 | 61 | |
GenBank mRNA | BC089994.1 | - | 4 - 5 | 551 | 7320068 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |