Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rltpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255548.2 | XP_006255610.1 | 24 - 25 | 2908 | 65 | |
XM_008772559.1 | XP_008770781.1 | 24 - 25 | 2908 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |