Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008722.1 | NP_001008722.1 | 3 - 4 | 448 | 74 | |
XM_006255737.2 | XP_006255799.1 | 4 - 5 | 496 | 74 | ||
GenBank mRNA | AB120371.1 | - | 3 - 4 | 448 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |