Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022399.2 | NP_071794.1 | 2 - 3 | 261 | 82 | |
GenBank mRNA | BC062395.1 | - | 2 - 3 | 238 | 5623178 | 82 |
BU671467.1 | - | 2 - 3 | 261 | 82 | ||
D78308.1 | - | 2 - 3 | 213 | 82 | ||
FQ210587.1 | - | 266 | 82 | |||
FQ220096.1 | - | 2 - 3 | 267 | 82 | ||
X53363.1 | - | 2 - 3 | 233 | 82 | ||
X79327.1 | - | 2 - 3 | 211 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |