Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5k1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042621.1 | NP_001036086.1 | 9 - 10 | 1506 | 74 | |
XM_008761315.1 | XP_008759537.1 | 9 - 10 | 1635 | 74 | ||
XM_008761316.1 | XP_008759538.1 | 9 - 10 | 1632 | 74 | ||
GenBank mRNA | AY724476.1 | - | 9 - 10 | 1506 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |