Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106449.2 | NP_001099919.1 | 3 - 4 | 447 | 69 | |
NM_001270650.1 | NP_001257579.1 | 4 - 5 | 521 | 69 | ||
NM_001270651.1 | NP_001257580.1 | 4 - 5 | 494 | 69 | ||
NM_001270652.1 | NP_001257581.1 | 3 - 4 | 424 | 69 | ||
NM_001270653.1 | NP_001257582.1 | 3 - 4 | 451 | 69 | ||
XM_006232855.2 | XP_006232917.1 | 4 - 5 | 516 | 69 | ||
XM_008761282.1 | XP_008759504.1 | 3 - 4 | 482 | 69 | ||
GenBank mRNA | FN800735.1 | - | 3 - 4 | 405 | 69 | |
FQ214903.1 | - | 3 - 4 | 384 | 69 | ||
FQ215988.1 | - | 4 - 5 | 450 | 69 | ||
FQ216125.1 | - | 3 - 4 | 373 | 69 | ||
FQ216236.1 | - | 1 - 2 | 118 | 69 | ||
FQ216249.1 | - | 4 - 5 | 444 | 69 | ||
FQ216534.1 | - | 3 - 4 | 350 | 69 | ||
FQ217203.1 | - | 3 - 4 | 371 | 69 | ||
FQ224853.1 | - | 3 - 4 | 372 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |