Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005761.1 | NP_001005761.1 | 3 - 4 | 656 | 93 | |
GenBank mRNA | AF526269.1 | - | 3 - 4 | 268 | 93 | |
BC083644.1 | - | 3 - 4 | 656 | 7134963 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |