Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025751.1 | NP_001020922.1 | 10 - 11 | 1359 | 61 | |
XM_006252188.2 | XP_006252250.1 | 11 - 12 | 1753 | 61 | ||
XM_008770769.1 | XP_008768991.1 | 10 - 11 | 1780 | 61 | ||
XM_008770770.1 | XP_008768992.1 | 9 - 10 | 1019 | 61 | ||
GenBank mRNA | BC098683.1 | - | 10 - 11 | 1359 | 7385175 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |