Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013085.1 | NP_001013103.1 | 4 - 5 | 283 | 89 | |
XM_006236486.2 | XP_006236548.1 | 4 - 5 | 319 | 89 | ||
XM_006236488.2 | XP_006236550.1 | 4 - 5 | 724 | 89 | ||
XM_006236489.2 | XP_006236551.1 | 4 - 5 | 590 | 89 | ||
XM_006236490.1 | XP_006236552.1 | 4 - 5 | 345 | 89 | ||
GenBank mRNA | BC091290.1 | - | 4 - 5 | 283 | 7306793 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |