Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248111.2 | XP_006248173.1 | 5 - 6 | 2475 | 86 | |
XM_006248112.2 | XP_006248174.1 | 5 - 6 | 2542 | 86 | ||
XM_006248114.2 | XP_006248176.1 | 4 - 5 | 2336 | 86 | ||
XM_008768611.1 | XP_008766833.1 | 5 - 6 | 1674 | 86 | ||
XM_008768612.1 | XP_008766834.1 | 4 - 5 | 1539 | 86 | ||
XM_221630.8 | XP_221630.6 | 4 - 5 | 1655 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |