Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107941.1 | NP_001101411.1 | 2 - 3 | 1799 | 75 | |
XM_006257515.2 | XP_006257577.1 | 2 - 3 | 1999 | 75 | ||
XM_008773519.1 | XP_008771741.1 | 2 - 3 | 1999 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |