Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blvrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106236.1 | NP_001099706.1 | 1 - 2 | 225 | 74 | |
GenBank mRNA | BC168943.1 | - | 1 - 2 | 129 | 7322093 | 74 |
FQ228106.1 | - | 1 - 2 | 109 | 74 | ||
FQ230737.1 | - | 1 - 2 | 172 | 74 | ||
FQ232598.1 | - | 1 - 2 | 172 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |