Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107498.1 | NP_001100968.1 | 3 - 4 | 1121 | 77 | |
XM_008759143.1 | XP_008757365.1 | 4 - 5 | 1244 | 77 | ||
GenBank mRNA | BC090319.1 | - | 4 - 5 | 1059 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |