Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Irf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006969.1 | NP_001006970.1 | 4 - 5 | 552 | 61 | |
XM_006229021.1 | XP_006229083.1 | 4 - 5 | 1154 | 61 | ||
XM_006229022.2 | XP_006229084.1 | 4 - 5 | 1114 | 61 | ||
XM_006229023.2 | XP_006229085.1 | 4 - 5 | 673 | 61 | ||
XM_006229024.2 | XP_006229086.1 | 4 - 5 | 541 | 61 | ||
XM_008759337.1 | XP_008757559.1 | 3 - 4 | 502 | 61 | ||
GenBank mRNA | BC083776.1 | - | 4 - 5 | 552 | 7193483 | 61 |
FQ213393.1 | - | 4 - 5 | 688 | 61 | ||
FQ218848.1 | - | 4 - 5 | 700 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |