Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748825.3 | XP_003748873.1 | 5 - 6 | 1003 | 60 | |
XM_006228804.2 | XP_006228866.1 | 4 - 5 | 1489 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |