Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spock3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107310.1 | NP_001100780.1 | 7 - 8 | 850 | 93 | |
XM_008771117.1 | XP_008769339.1 | 8 - 9 | 841 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |