Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stoml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031646.1 | NP_001026816.1 | 7 - 8 | 726 | 94 | |
GenBank mRNA | BC099164.1 | - | 7 - 8 | 726 | 7382023 | 94 |
FQ213146.1 | - | 7 - 8 | 757 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |