Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238095.2 | XP_006238157.1 | 1 - 2 | 931 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |