Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3gl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053935.1 | NP_446387.1 | 2 - 3 | 197 | 61 | |
XM_006238359.2 | XP_006238421.1 | 2 - 3 | 1581 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |