Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004071.1 | NP_001004071.1 | 11 - 12 | 1219 | 78 | |
XM_008767382.1 | XP_008765604.1 | 12 - 13 | 2873 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |