Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133412.1 | NP_596903.1 | 11 - 12 | 2076 | 132 | |
GenBank mRNA | AF321415.1 | - | 11 - 12 | 2076 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |