Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108852.2 | NP_001102322.1 | 2 - 3 | 269 | 81 | |
GenBank mRNA | BC162041.1 | - | 2 - 3 | 269 | 7100495 | 81 |
FQ226241.1 | - | 2 - 3 | 288 | 81 | ||
FQ229096.1 | - | 2 - 3 | 314 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |